3-Amino-5-bromo-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-1H-indazol-3-amine
A474142-1G
A474142-25G
A474142-5G
A474142-100G
A474142-250MG
A474142-10G
103828-058EA
9
USD
103828-058
103839-132
76843-056
76843-058
76843-060
77169-052
77169-054
77572-492
3-Amino-5-bromo-1H-indazole 97%
3-Amino-5-bromo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₆BrN₃ MW: 212.05 g/mol Boiling Pt: 431.3 °C (760 mm Hg) Melting Pt: 171…175 °C Flash Pt: 214.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426696 CAS Number: 61272-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...