5-Bromo-1H-imidazole-4-carbaldehyde ≥95%

Supplier: Ambeed
Warning

A147390-250MG A147390-100MG
103803-768EA 257 USD
103803-768 77592-764 77683-256
5-Bromo-1H-imidazole-4-carbaldehyde ≥95%
5-Bromo-1H-imidazole-4-carbaldehyde

Precaución: For research use only.

Formula: C₄H₃BrN₂O
MW: 174.983 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21336623
CAS Number: 50743-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR