5-Bromo-1H-imidazo[4,5-b]pyridine, 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3H-imidazo[4,5-b]pyridine, 5-Bromo-4-azabenzimidazole, 5-Bromo-1H-imidazo[4,5-b]pyridine

A151511-250MG A151511-1G A151511-5G
103804-264EA 74 USD
103804-264 103842-416 76783-388 77317-986 77317-988
5-Bromo-1H-imidazo[4,5-b]pyridine, 95%
5-Bromo-1H-imidazo[4,5-b]pyridine,

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
Boiling Pt: 442.5 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11227145
CAS Number: 28279-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR