5-Bromo-1H-benzoimidazole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A194428-1G A194428-250MG
103816-654EA 45 USD
103816-654 103847-264 77085-182 77164-368
5-Bromo-1H-benzoimidazole-2-carboxylic acid 97%
5-Bromo-1H-benzoimidazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
Flash Pt: 252.944 °C
Storage Temperature: Freezer
MDL Number: MFCD06738769
CAS Number: 40197-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR