5-Bromo-1,3-dimethyluracil 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine, 5-Bromo-1,3-dimethylpyrimidine-2,4(1H,3H)-dione

A101401-250MG A101401-25G
103795-956EA 33 USD
103795-956 103821-640 76765-166 76765-168
5-Bromo-1,3-dimethyluracil 95%
5-Bromo-1,3-dimethyluracil

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.036 g/mol
Boiling Pt: 242.1 °C (760 mm Hg)
Melting Pt: 182…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191846
CAS Number: 7033-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR