5-Bromo-1,3-dimethyl-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
A184477-100MG
A184477-250MG
A184477-5G
103815-754EA
49
USD
103815-754
103846-318
77012-078
77012-080
77083-932
5-Bromo-1,3-dimethyl-1H-pyrazole 97%
5-Bromo-1,3-dimethyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₅H₇BrN₂ Boiling Pt: 211.7 °C (760 mm Hg) Flash Pt: 81.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08448181 CAS Number: 5744-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...