5-Bromo-1,3-dihydrobenzo[c]thiophene 2,2-dioxide 97%

Supplier: Ambeed
Warning

A130765-100MG A130765-250MG A130765-25MG A130765-50MG A130765-5G
103801-478EA 618 USD
103801-478 77149-604 77149-606 77149-608 77149-610 77313-946
5-Bromo-1,3-dihydrobenzo[c]thiophene 2,2-dioxide 97%
5-Bromo-1,3-dihydrobenzo[c]thiophene 2,2-dioxide

Precaución: for Research Use Only

Formula: C₈H₇BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD00666887
CAS Number: 351005-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR