5-Bromo-1,3-difluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoro-4-nitrobromobenzene

A197916-100G A197916-10G A197916-500G A197916-1G A197916-25G A197916-5G
103816-976EA 10 USD
103816-976 103847-600 76975-138 76975-140 76975-142 77085-610 77085-612 77085-614
5-Bromo-1,3-difluoro-2-nitrobenzene 98%
5-Bromo-1,3-difluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MW: 237.9864 g/mol
Boiling Pt: 265.7 °C (760 mm Hg)
Flash Pt: 114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12025434
CAS Number: 147808-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR