1-Bromo-3,5-difluoro-4-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1,3-difluoro-2-iodobenzene

A179331-100G A179331-10G A179331-500G A179331-25G A179331-5G
103845-764EA 8 USD
103845-764 76998-604 76998-606 76998-608 77083-220 77083-222
1-Bromo-3,5-difluoro-4-iodobenzene 98%
1-Bromo-3,5-difluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂I
MW: 318.8854 g/mol
Boiling Pt: 236 °C (760 mm Hg)
Melting Pt: 40…41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861126
CAS Number: 160976-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR