5-Bromo-1,3-dichloro-2-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-4-fluorobromobenzene

A144229-100G A144229-25G A144229-500G A144229-1G A144229-5G A144229-10G
103803-382EA 9 USD
103803-382 103841-458 76783-086 76783-088 76783-090 77316-696 77316-698 77667-080
5-Bromo-1,3-dichloro-2-fluorobenzene 98%
5-Bromo-1,3-dichloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
MW: 243.8900 g/mol
Boiling Pt: 234.3 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09996901
CAS Number: 17318-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR