5-Bromo-1,2,3,4-tetrahydroquinoline 97%
Supplier: Ambeed
|
Warning
|
A200491-10G
A200491-250MG
A200491-1G
A200491-5G
A200491-25G
103817-208EA
31
USD
103817-208
103847-836
76814-276
76814-278
77085-896
77085-898
77302-688
5-Bromo-1,2,3,4-tetrahydroquinoline 97%
5-Bromo-1,2,3,4-tetrahydroquinoline
Precaución: for Research Use Only
Formula:
C₉H₁₀BrN MW: 212.09 g/mol Boiling Pt: 299.676 °C (760 mm Hg) Flash Pt: 135 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08544260 CAS Number: 114744-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...