5-Bromo-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: Ambeed
Warning

A189602-1G A189602-5G A189602-10G A189602-250MG
103816-208EA 31 USD
103816-208 103846-802 77084-576 77084-578 77111-706 77111-708
5-Bromo-1,2,3,4-tetrahydroisoquinoline 97%
5-Bromo-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀BrN
MW: 212.0900 g/mol
Boiling Pt: 294.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08544279
CAS Number: 81237-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR