5-Bromo-1,2,3,4-tetrahydroisoquinolin-8-amine 98%

Supplier: Ambeed
Warning

A296292-100MG A296292-250MG
103814-090EA 69 USD
103814-090 103835-230 76875-592 76875-594
5-Bromo-1,2,3,4-tetrahydroisoquinolin-8-amine 98%
5-Bromo-1,2,3,4-tetrahydroisoquinolin-8-amine

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂
MW: 227.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193651
CAS Number: 1260779-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR