5-Bromo-1,10-phenanthroline 97%

Supplier: Ambeed
Warning

A261897-250MG A261897-1G A261897-25G A261897-5G A261897-10G
103812-544EA 29 USD
103812-544 103852-434 76771-896 77092-046 77092-048 77092-050 77601-116
5-Bromo-1,10-phenanthroline 97%
5-Bromo-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₂H₇BrN₂
Boiling Pt: 417.6 °C (760 mm Hg)
Flash Pt: 206.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00046903
CAS Number: 40000-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR