5'-Bromo-m-terphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,5-diphenylbenzene, 5'-Bromo-1,1':3',1''-terphenyl

A121640-100G A121640-10G A121640-250MG A121640-25G A121640-500G A121640-1G A121640-5G
103800-102EA 6 USD
103800-102 76822-462 76822-464 76822-466 76822-468 76822-470 77312-226 77312-228
5'-Bromo-m-terphenyl 98%
5'-Bromo-m-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₃Br
MW: 309.21 g/mol
Boiling Pt: 401.1 °C (760 mm Hg)
Melting Pt: 105…106 °C
Flash Pt: 192.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00196170
CAS Number: 103068-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR