5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 98%

Supplier: Ambeed
Warning

A207241-100G A207241-100MG A207241-10G A207241-250MG A207241-25G A207241-1G A207241-5G
103817-776EA 9 USD
103817-776 103848-456 77030-338 77030-340 77030-342 77030-344 77030-346 77086-724 77086-726
5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 98%
5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₅BrN₄
MW: 213.0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11655928
CAS Number: 1010120-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR