5-Boc-hexahydropyrrolo[3,4-b]pyrrole 97%
Supplier: Ambeed
|
Warning
|
A321330-100MG
A321330-10G
A321330-250MG
A321330-1G
A321330-5G
103825-268EA
55
USD
103825-268
103836-202
77114-726
77114-728
77114-730
77306-236
77306-238
5-Boc-hexahydropyrrolo[3,4-b]pyrrole 97%
5-Boc-hexahydropyrrolo[3,4-b]pyrrole
Precaución: for Research Use Only
Formula:
C₁₁H₂₀N₂O₂ MW: 212.2887 g/mol Boiling Pt: 295.4 °C (760 mm Hg) Flash Pt: 132.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08235016 CAS Number: 132414-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...