5-Azaindole 98%

Supplier: Ambeed
Warning

Synonyms: 1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine

A143044-10G A143044-250MG A143044-1G A143044-25G A143044-5G A143044-100G
103803-234EA 8 USD
103803-234 77137-398 77137-400 77316-480 77316-482 77316-484 77566-868
5-Azaindole 98%
5-Azaindole

Precaución: For research use only.

Formula: C₇H₆N₂
Boiling Pt: 295.8 °C (760 mm Hg)
Melting Pt: 104.6…108.4 °C
Flash Pt: 137.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00955936
CAS Number: 271-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR