5-Aminothiazole-4-carboxamide 98%

Supplier: Ambeed
Warning

A336425-100MG A336425-250MG
103825-716EA 78 USD
103825-716 103836-690 76876-088 76876-090
5-Aminothiazole-4-carboxamide 98%
5-Aminothiazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₄H₅N₃OS
Storage Temperature: Refrigerator
MDL Number: MFCD11848433
CAS Number: 5539-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR