5-Aminopyrimidine 98%

Supplier: Ambeed
Warning

A392008-1G A392008-25G A392008-10G A392008-250MG
103826-892EA 18 USD
103826-892 103837-930 76841-324 76841-326 77168-228 77168-230
5-Aminopyrimidine 98%
5-Aminopyrimidine

Precaución: for Research Use Only

Formula: C₄H₅N₃
MW: 95.1000 g/mol
Boiling Pt: 240.7 °C (760 mm Hg)
Melting Pt: 170…174 °C
Flash Pt: 122.563 °C
Storage Temperature: Ambient
MDL Number: MFCD01529870
CAS Number: 591-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR