5-Aminopyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A269747-100MG A269747-250MG A269747-25G
103812-976EA 27 USD
103812-976 103852-882 77031-884 77031-886 77872-311
5-Aminopyrimidin-4(3H)-one 97%
5-Aminopyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₄H₅N₃O
Melting Pt: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD11574230
CAS Number: 69785-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR