5-Aminopyridine-2-carboxamide 97%

Supplier: Ambeed
Warning

A184798-250MG A184798-1G A184798-5G
103815-786EA 79 USD
103815-786 103846-352 76769-696 77301-916 77301-918
5-Aminopyridine-2-carboxamide 97%
5-Aminopyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
MW: 137.1393 g/mol
Boiling Pt: 427.3 °C (760 mm Hg)
Flash Pt: 212.23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09879701
CAS Number: 145255-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR