5-Aminopyridin-3-ol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-5-hydroxypyridine
A197132-1G
A197132-5G
A197132-100MG
A197132-250MG
103816-896EA
95
USD
103816-896
103847-514
77085-496
77085-498
77274-670
77274-672
5-Aminopyridin-3-ol 97%
5-Aminopyridin-3-ol
Precaución: for Research Use Only
Formula:
C₅H₆N₂O MW: 110.1139 g/mol Boiling Pt: 447.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD04114213 CAS Number: 3543-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...