2-Amino-5-cyanopyrazine 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Aminopyrazine-2-carbonitrile
A120830-10G
A120830-250MG
A120830-1G
A120830-5G
A120830-25G
103799-996EA
16
USD
103799-996
77009-428
77009-430
77297-790
77297-792
77683-085
2-Amino-5-cyanopyrazine 98%, Ambeed.Inc
2-Amino-5-cyanopyrazine
Precaución: For research use only.
Formula:
C₅H₄N₄ MW: 120.1100 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08234954 CAS Number: 113305-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...