5-Aminopicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid

A173814-1G A173814-25G A173814-5G A173814-10G A173814-250MG
103806-874EA 10 USD
103806-874 103845-172 77082-454 77082-456 77082-458 77111-126 77111-128
5-Aminopicolinic acid 98%
5-Aminopicolinic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 420 °C (760 mm Hg)
Melting Pt: 222…226 °C
Flash Pt: 208.3 °C
Storage Temperature: Ambient
MDL Number: MFCD02684600
CAS Number: 24242-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR