5-Aminonicotinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-3-pyridinecarbonitrile

A231878-100MG A231878-250MG A231878-1G A231878-5G
103810-798EA 26 USD
103810-798 103850-552 76771-178 76771-180 77089-452 77089-454
5-Aminonicotinonitrile 97%
5-Aminonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.1200 g/mol
Boiling Pt: 349.3 °C (760 mm Hg)
Melting Pt: 118…123 °C
Flash Pt: 165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367907
CAS Number: 13600-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR