5-Amino-1H-indazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine
A177014-1G
A177014-25G
A177014-100G
A177014-10G
A177014-500G
A177014-5G
103807-200EA
5
USD
103807-200
103845-526
77082-902
77082-904
77138-578
77138-580
77138-582
77301-484
5-Amino-1H-indazole 98%
5-Amino-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₇N₃ MW: 133.15 g/mol Boiling Pt: 376.6 °C (760 mm Hg) Melting Pt: 175…178 °C Flash Pt: 209.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037975 CAS Number: 19335-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...