5-Aminofluorescein (isomer I) 96%
Supplier: Ambeed
|
Warning
|
A129725-250MG
A129725-1G
A129725-5G
103801-316EA
57
USD
103801-316
76797-818
77298-364
77298-366
5-Aminofluorescein (isomer I) 96%
5-Aminofluorescein (isomer I)
Precaución: for Research Use Only
Formula:
C₂₀H₁₃NO₅ Boiling Pt: 695.3 °C (760 mm Hg) Melting Pt: 223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005052 CAS Number: 3326-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...