5-Aminobenzo[b]thiophene 95%

Supplier: Ambeed
Warning

A111437-10G A111437-5G A111437-250MG A111437-25G
103797-832EA 72 USD
103797-832 103823-652 76687-606 77284-110 77570-644 77876-830
5-Aminobenzo[b]thiophene 95%
5-Aminobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₇NS
Boiling Pt: 369.5 °C (760 mm Hg)
Melting Pt: 72…75 °C
Flash Pt: 195.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04115381
CAS Number: 20532-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR