5-Amino-7-azaindole 97%

Supplier: Ambeed
Warning

A299680-10G A299680-25G A299680-1G A299680-5G A299680-250MG
103814-234EA 29 USD
103814-234 103835-384 77014-922 77014-924 77305-774 77305-776 77890-568
5-Amino-7-azaindole 97%
5-Amino-7-azaindole

Precaución: for Research Use Only

Formula: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD06659683
CAS Number: 100960-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR