5-Amino-6-methylpyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-6-methylnicotinonitrile

A285388-100MG A285388-10G A285388-250MG
103813-676EA 358 USD
103813-676 103834-810 77014-660 77014-662 77014-664
5-Amino-6-methylpyridine-3-carbonitrile 97%
5-Amino-6-methylpyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13175887
CAS Number: 3308-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR