5-Amino-6-methyl-1,3-dihydro-benzoimidazol-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-6-methyl-1H-benzo[d]imidazol-2(3H)-one, 5-Amino-6-methylbenzimidazolone

A287574-10G A287574-250MG A287574-1G A287574-5G A287574-25G
103813-764EA 34 USD
103813-764 103834-906 76816-504 76816-506 76837-460 76837-462 77305-518
5-Amino-6-methyl-1,3-dihydro-benzoimidazol-2-one 95%
5-Amino-6-methyl-1,3-dihydro-benzoimidazol-2-one

Precaución: for Research Use Only

Formula: C₈H₉N₃O
Boiling Pt: 215 °C (760 mm Hg)
Melting Pt: 170…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684428
CAS Number: 67014-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR