5-Amino-3-methyl-1,2,4-thiadiazole 98%

Supplier: Ambeed
Warning

A182705-100G A182705-10G A182705-25G A182705-5G A182705-1G
103807-740EA 6 USD
103807-740 103846-116 76974-504 76974-506 77083-666 77083-668 77301-800
5-Amino-3-methyl-1,2,4-thiadiazole 98%
5-Amino-3-methyl-1,2,4-thiadiazole

Precaución: for Research Use Only

Formula: C₃H₅N₃S
Boiling Pt: 261.2 °C (760 mm Hg)
Melting Pt: 202 °C
Flash Pt: 111.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01114751
CAS Number: 17467-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR