3-Amino-5-bromobenzonitrile 95%

Supplier: Ambeed
Warning

A240467-100MG A240467-10G A240467-250MG A240467-1G
103811-348EA 13 USD
103811-348 103851-150 77031-268 77031-270 77031-272 77090-302
3-Amino-5-bromobenzonitrile 95%
3-Amino-5-bromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Flash Pt: 147.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11109874
CAS Number: 49674-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR