5-Amino-3-(trifluoromethyl)picolinonitrile 97%
Supplier: Ambeed
|
Warning
|
A345390-1G
A345390-5G
A345390-100G
A345390-100MG
A345390-250MG
A345390-25G
A345390-10G
103825-922EA
13
USD
103825-922
103836-904
76839-962
76839-964
77015-640
77015-642
77015-644
77015-646
77612-974
5-Amino-3-(trifluoromethyl)picolinonitrile 97%
5-Amino-3-(trifluoromethyl)picolinonitrile
Precaución: for Research Use Only
Formula:
C₇H₄F₃N₃ MW: 187.122 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17010171 CAS Number: 573762-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...