5-Amino-2-methylthiazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A226435-1G A226435-100MG A226435-250MG
103819-246EA 108 USD
103819-246 103850-148 77088-878 77193-008 77193-010
5-Amino-2-methylthiazole-4-carboxylic acid 97%
5-Amino-2-methylthiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MW: 158.18 g/mol
Boiling Pt: 363.1 °C (760 mm Hg)
Flash Pt: 173.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12546655
CAS Number: 36405-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR