5-Amino-2-methylphenol 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-o-cresol, 3-Hydroxy-4-methylaniline, 1-Amino-3-hydroxy-4-methylbenzene, 5-Amino-2-methylphenol, 4-Amino-2-hydroxytoluene

A165259-100G A165259-500G A165259-25G
103808-956EA 179 USD
103808-956 76768-780 76768-782 77320-410
5-Amino-2-methylphenol 95%
5-Amino-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 268.6 °C (760 mm Hg)
Melting Pt: 160…162 °C
Flash Pt: 116.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00043922
CAS Number: 2835-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR