5-Amino-2-methylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-methylbenzoic Acid, 2-Methyl-5-aminobenzoic acid, 5-Amino-o-toluic acid

A101203-100G A101203-10G
103821-540EA 7 USD
103821-540 77270-208 77270-210
5-Amino-2-methylbenzoic acid 97%
5-Amino-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 348.8 °C (760 mm Hg)
Melting Pt: 194…199 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06208351
CAS Number: 2840-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR