5-Amino-2-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A136098-250MG A136098-1G A136098-5G
103802-274EA 22 USD
103802-274 77258-656 77315-168 77315-170
5-Amino-2-methoxypyrimidine 98%
5-Amino-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.1286 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234878
CAS Number: 56621-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR