5-Amino-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A221563-25G A221563-100G A221563-10G A221563-500G A221563-5G
103849-814EA 7 USD
103849-814 77088-412 77112-772 77112-774 77112-776 77303-606
5-Amino-2-methoxypyridine 98%
5-Amino-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 264.8 °C (760 mm Hg)
Melting Pt: 29…31 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006264
CAS Number: 6628-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR