5-Amino-2-methoxybenzonitrile 95%

Supplier: Ambeed
Warning

A113814-10G A113814-250MG A113814-1G A113814-5G A113814-25G
103798-296EA 55 USD
103798-296 103824-140 77025-924 77025-926 77310-958 77310-960 77871-467
5-Amino-2-methoxybenzonitrile 95%
5-Amino-2-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.1619 g/mol
Flash Pt: 163.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00723841
CAS Number: 214623-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR