5-Amino-2-hydroxybenzonitrile 95%

Supplier: Ambeed
Warning

A189610-100MG A189610-250MG A189610-1G
103816-212EA 164 USD
103816-212 103846-806 76769-848 76769-850 77084-580
5-Amino-2-hydroxybenzonitrile 95%
5-Amino-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646169
CAS Number: 87029-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR