5-Amino-2-fluoropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-pyridylamine, 2-Fluoropyridin-5-amine

A144459-100G A144459-10G A144459-500G A144459-1G A144459-25G A144459-5G
103803-408EA 5 USD
103803-408 103841-484 77285-874 77285-876 77285-878 77299-386 77316-728 77316-730
5-Amino-2-fluoropyridine 98%
5-Amino-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅FN₂
MW: 112.11 g/mol
Boiling Pt: 264 °C (760 mm Hg)
Melting Pt: 86…87 °C
Flash Pt: 113.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01632180
CAS Number: 1827-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR