5-Amino-2-fluoroisonicotinic acid 97%

Supplier: Ambeed
Warning

A221973-100MG A221973-250MG A221973-1G A221973-5G
103818-958EA 56 USD
103818-958 103849-834 76999-954 76999-956 77088-426 77088-428
5-Amino-2-fluoroisonicotinic acid 97%
5-Amino-2-fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
Boiling Pt: 514.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03411724
CAS Number: 171178-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR