5-Amino-2-chloropyrimidine 98%
Supplier: Ambeed
|
Warning
|
A200494-250MG
A200494-1G
A200494-5G
A200494-25G
A200494-100G
103817-210EA
23
USD
103817-210
103847-838
76814-280
77085-900
77085-902
77567-898
77883-457
5-Amino-2-chloropyrimidine 98%
5-Amino-2-chloropyrimidine
Precaución: For research use only.
Formula:
C₄H₄ClN₃ Boiling Pt: 338.8 °C (760 mm Hg) Melting Pt: 196…200 °C Flash Pt: 158.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06412560 CAS Number: 56621-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...