5-Amino-2-chloropyridine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-amino-2-chloropyridine-4-carboxylic acid, 5-Amino-2-chloroisonicotinic acid, 3-Amino-6-chloroisonicotinic acid

A268520-100G A268520-10G A268520-250MG A268520-1G A268520-25G A268520-5G
103812-918EA 8 USD
103812-918 103852-820 76816-146 76816-148 76816-150 77092-572 77092-574 77092-576
5-Amino-2-chloropyridine-4-carboxylic acid 98%
5-Amino-2-chloropyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
Flash Pt: 264.906 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659499
CAS Number: 58483-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR