5-Amino-2-chlorobenzothiazole 98%

Supplier: Ambeed
Warning

A205593-100MG A205593-250MG A205593-1G A205593-5G
103817-644EA 49 USD
103817-644 103848-310 76785-478 76785-480 77086-532 77086-534
5-Amino-2-chlorobenzothiazole 98%
5-Amino-2-chlorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅ClN₂S
MW: 184.6460 g/mol
Melting Pt: 55…59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11040282
CAS Number: 80945-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR