5-Amino-2-chlorobenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-cyanoaniline

A345553-10G A345553-250MG A345553-25G A345553-5G A345553-1G
103825-928EA 16 USD
103825-928 103836-910 76773-230 76773-232 76839-974 76839-976 77306-568
5-Amino-2-chlorobenzonitrile 97%
5-Amino-2-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
Boiling Pt: 339.1 °C (760 mm Hg)
Flash Pt: 158.913 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00800459
CAS Number: 35747-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR