5-Amino-2-bromopyrimidine 95%
Supplier: Ambeed
|
Warning
|
A298486-1G
A298486-5G
A298486-100MG
A298486-250MG
103814-184EA
63
USD
103814-184
103835-328
76838-024
76838-026
77262-950
77262-952
5-Amino-2-bromopyrimidine 95%
5-Amino-2-bromopyrimidine
Precaución: for Research Use Only
Formula:
C₄H₄BrN₃ Boiling Pt: 364.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD07778608 CAS Number: 56621-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...