Luminol (3-Aminophthalhydrazide) 98%

Supplier: Ambeed
Warning

Synonyms: NSC 5064, Luminol, 5-Amino-2,3-Dihydro-1,4-Phthalazinedione, 5-Amino-2,3-dihydrophthalazine-1,4-dione

A166579-100MG A166579-10G A166579-25G A166579-5G A166579-100G A166579-500G
103844-342EA 7 USD
103844-342 77236-138 77236-140 77320-604 77320-606 77615-384 77746-123
Luminol (3-Aminophthalhydrazide) 98%
Luminol (3-Aminophthalhydrazide)

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Boiling Pt: 621.9 °C (760 mm Hg)
Melting Pt: 325…330 °C
Flash Pt: 329.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006890
CAS Number: 521-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR